siRNA Details
| Gene Symbol: Slc30a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130538.1 | NP_001124010.1 | 6 | 1070 | ||
| XM_006241645.2 | XP_006241707.1 | 6 | 1070 | |||
| XM_008765500.1 | XP_008763722.1 | 6 | 827 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a8| Gene Symbol: Slc30a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130538.1 | NP_001124010.1 | 6 | 1070 | ||
| XM_006241645.2 | XP_006241707.1 | 6 | 1070 | |||
| XM_008765500.1 | XP_008763722.1 | 6 | 827 | |||