siRNA Details
Gene Symbol: Gsdmc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134495.1 | NP_001127967.1 | 4 | 597 | ||
XM_006241631.2 | XP_006241693.1 | 7 | 1237 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gsdmc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134495.1 | NP_001127967.1 | 4 | 597 | ||
XM_006241631.2 | XP_006241693.1 | 7 | 1237 |