siRNA Details
| Gene Symbol: Fam49b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126267.2 | NP_001119739.1 | 8, 9 | 753 | ||
| XM_006241632.2 | XP_006241694.1 | 6, 7 | 1865 | |||
| XM_006241633.1 | XP_006241695.1 | 7, 8 | 744 | |||
| XM_006241634.1 | XP_006241696.1 | 6, 7 | 603 | |||
| XM_006241635.2 | XP_006241697.1 | 7, 8 | 724 | |||
| XM_008765501.1 | XP_008763723.1 | 8, 9 | 832 | |||
| GenBank mRNA | BC166469.1 | - | 8, 9 | 753 | 9036548 | |
| FQ212269.1 | - | 7, 8 | 749 | |||

Fam49b
Genomic Map