siRNA Details
| Gene Symbol: Nsmce2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024876.1 | NP_001020047.1 | 6 | 662 | ||
| XM_008765505.1 | XP_008763727.1 | 5 | 2822 | |||
| GenBank mRNA | BC097477.1 | - | 6 | 662 | 7458086 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nsmce2| Gene Symbol: Nsmce2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024876.1 | NP_001020047.1 | 6 | 662 | ||
| XM_008765505.1 | XP_008763727.1 | 5 | 2822 | |||
| GenBank mRNA | BC097477.1 | - | 6 | 662 | 7458086 | |