siRNA Details
| Gene Symbol: Atp6v1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011992.1 | NP_001011992.1 | 11 | 969 | ||
| XM_006241602.2 | XP_006241664.1 | 10 | 974 | |||
| GenBank mRNA | BC089961.1 | - | 11 | 969 | 7314200 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v1c1| Gene Symbol: Atp6v1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011992.1 | NP_001011992.1 | 11 | 969 | ||
| XM_006241602.2 | XP_006241664.1 | 10 | 974 | |||
| GenBank mRNA | BC089961.1 | - | 11 | 969 | 7314200 | |