siRNA Details
Gene Symbol: Grhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134527.1 | NP_001127999.1 | 12, 13 | 1657 | ||
GenBank mRNA | BC168688.1 | - | 12, 13 | 1657 | 7932577 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Grhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134527.1 | NP_001127999.1 | 12, 13 | 1657 | ||
GenBank mRNA | BC168688.1 | - | 12, 13 | 1657 | 7932577 |