siRNA Details
| Gene Symbol: Grhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134527.1 | NP_001127999.1 | 4, 5 | 821 | ||
| XM_006241552.2 | XP_006241614.1 | 4, 5 | 1649 | |||
| GenBank mRNA | BC168688.1 | - | 4, 5 | 821 | 7932577 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grhl2| Gene Symbol: Grhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134527.1 | NP_001127999.1 | 4, 5 | 821 | ||
| XM_006241552.2 | XP_006241614.1 | 4, 5 | 1649 | |||
| GenBank mRNA | BC168688.1 | - | 4, 5 | 821 | 7932577 | |