siRNA Details
| Gene Symbol: Top1mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002798.1 | NP_001002798.1 | 6 | 790 | ||
| XM_006241784.2 | XP_006241846.1 | 6 | 990 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Top1mt| Gene Symbol: Top1mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002798.1 | NP_001002798.1 | 6 | 790 | ||
| XM_006241784.2 | XP_006241846.1 | 6 | 990 | |||