siRNA Details
| Gene Symbol: LOC102548778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241910.2 | XP_006241972.1 | 5 | 997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102548778| Gene Symbol: LOC102548778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241910.2 | XP_006241972.1 | 5 | 997 | ||