siRNA Details
| Gene Symbol: Foxh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130493.1 | NP_001123965.1 | 3 | 1199 | ||
| XM_008765565.1 | XP_008763787.1 | 3 | 1436 | |||
| GenBank mRNA | BC158731.1 | - | 3 | 1109 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxh1| Gene Symbol: Foxh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130493.1 | NP_001123965.1 | 3 | 1199 | ||
| XM_008765565.1 | XP_008763787.1 | 3 | 1436 | |||
| GenBank mRNA | BC158731.1 | - | 3 | 1109 | ||