siRNA Details
| Gene Symbol: Recql4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130494.1 | NP_001123966.1 | 4 | 337 | ||
| XM_006241806.2 | XP_006241868.1 | 4 | 409 | |||
| XM_008765566.1 | XP_008763788.1 | 4 | 409 | |||
| XM_008765567.1 | XP_008763789.1 | 4 | 409 | |||
| XM_008765568.1 | XP_008763790.1 | 4 | 409 | |||

Recql4
Genomic Map