siRNA Details
| Gene Symbol: Gtpbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199315.1 | NP_001186244.1 | 4 | 847 | ||
| GenBank mRNA | GU256773.1 | - | 4 | 807 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtpbp1| Gene Symbol: Gtpbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199315.1 | NP_001186244.1 | 4 | 847 | ||
| GenBank mRNA | GU256773.1 | - | 4 | 807 | ||