siRNA Details
Gene Symbol: Polr3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127295.2 | NP_001120767.1 | 3 | 206 | ||
GenBank mRNA | BC167088.1 | - | 3 | 206 | 5621972 | |
FQ220736.1 | - | 3 | 240 | |||
FQ225628.1 | - | 3 | 206 | |||
FQ229831.1 | - | 3 | 241 |
