siRNA Details
| Gene Symbol: Pnpla5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130497.1 | NP_001123969.1 | 6 | 862 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnpla5| Gene Symbol: Pnpla5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130497.1 | NP_001123969.1 | 6 | 862 | ||