siRNA Details
| Gene Symbol: Samm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004241.1 | NP_001004241.1 | 2, 3 | 198 | ||
| GenBank mRNA | BC079302.1 | - | 2, 3 | 198 | 7134547 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samm50| Gene Symbol: Samm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004241.1 | NP_001004241.1 | 2, 3 | 198 | ||
| GenBank mRNA | BC079302.1 | - | 2, 3 | 198 | 7134547 | |