siRNA Details
| Gene Symbol: Ribc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013949.1 | NP_001013971.2 | 1 | 261 | ||
| GenBank mRNA | BC079434.1 | - | 1 | 261 | 7115185 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ribc2| Gene Symbol: Ribc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013949.1 | NP_001013971.2 | 1 | 261 | ||
| GenBank mRNA | BC079434.1 | - | 1 | 261 | 7115185 | |