siRNA Details
| Gene Symbol: Celsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242167.2 | XP_006242229.1 | 25 | 8040 | ||
| XM_008765820.1 | XP_008764042.1 | 25 | 8041 | |||
| XM_008765821.1 | XP_008764043.1 | 25 | 8040 | |||
More Information
Set Membership: |
|

Celsr1
Genomic Map