siRNA Details
| Gene Symbol: Kif21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765827.1 | XP_008764049.1 | 36 | 4781 | ||
| XM_008765828.1 | XP_008764050.1 | 35 | 4766 | |||
| XM_008765829.1 | XP_008764051.1 | 33 | 4688 | |||
| XM_008765830.1 | XP_008764052.1 | 32 | 4667 | |||
| XM_008765831.1 | XP_008764053.1 | 33 | 4598 | |||
| XM_008765832.1 | XP_008764054.1 | 32 | 4559 | |||
| XM_008765833.1 | XP_008764055.1 | 32 | 4490 | |||
| XM_008765834.1 | XP_008764056.1 | 31 | 4469 | |||
| XM_008765835.1 | XP_008764057.1 | 31 | 4451 | |||
| XM_008765836.1 | XP_008764058.1 | 34 | 4766 | |||

Kif21a
Genomic Map