siRNA Details
| Gene Symbol: Ddx23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106793.2 | NP_001100263.2 | 12 | 1532 | ||
| XM_006257343.2 | XP_006257405.1 | 12 | 1621 | |||
| GenBank mRNA | BC169082.1 | - | 11 | 1411 | ||
| CO570626.1 | - | 2 | 166 | |||

Ddx23
Genomic Map