siRNA Details
| Gene Symbol: Myg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005545.1 | NP_001005545.1 | 7 | 1117 | ||
| GenBank mRNA | BC082112.1 | - | 7 | 1117 | 7135321 | |
| FQ213994.1 | - | 5 | 1182 | |||
| FQ230067.1 | - | 7 | 1147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myg1| Gene Symbol: Myg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005545.1 | NP_001005545.1 | 7 | 1117 | ||
| GenBank mRNA | BC082112.1 | - | 7 | 1117 | 7135321 | |
| FQ213994.1 | - | 5 | 1182 | |||
| FQ230067.1 | - | 7 | 1147 | |||