siRNA Details
| Gene Symbol: Ccdc82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007660.1 | NP_001007661.1 | 3 | 447 | ||
| XM_006242519.2 | XP_006242581.1 | 4 | 509 | |||
| GenBank mRNA | BC081989.1 | - | 3 | 447 | 7129126 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc82| Gene Symbol: Ccdc82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007660.1 | NP_001007661.1 | 3 | 447 | ||
| XM_006242519.2 | XP_006242581.1 | 4 | 509 | |||
| GenBank mRNA | BC081989.1 | - | 3 | 447 | 7129126 | |