siRNA Details
| Gene Symbol: Ldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175762.2 | NP_786938.1 | 17 | 2711 | ||
| GenBank mRNA | X13722.1 | - | 17 | 2711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ldlr| Gene Symbol: Ldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175762.2 | NP_786938.1 | 17 | 2711 | ||
| GenBank mRNA | X13722.1 | - | 17 | 2711 | ||