siRNA Details
| Gene Symbol: Thyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007661.2 | NP_001007662.1 | 3 | 310 | ||
| GenBank mRNA | BC064031.1 | - | 3 | 314 | 6887845 | |
| FQ213324.1 | - | 2 | 459 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thyn1| Gene Symbol: Thyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007661.2 | NP_001007662.1 | 3 | 310 | ||
| GenBank mRNA | BC064031.1 | - | 3 | 314 | 6887845 | |
| FQ213324.1 | - | 2 | 459 | |||