siRNA Details
| Gene Symbol: Vps26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106809.1 | NP_001100279.1 | 5 | 1180 | ||
| GenBank mRNA | BC161865.1 | - | 5 | 1116 | 7105156 | |
| CO394403.1 | - | 2 | 210 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps26b| Gene Symbol: Vps26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106809.1 | NP_001100279.1 | 5 | 1180 | ||
| GenBank mRNA | BC161865.1 | - | 5 | 1116 | 7105156 | |
| CO394403.1 | - | 2 | 210 | |||