siRNA Details
| Gene Symbol: Hepacam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729891.4 | XP_002729937.2 | 4 | 964 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hepacam| Gene Symbol: Hepacam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729891.4 | XP_002729937.2 | 4 | 964 | ||