siRNA Details
| Gene Symbol: Esam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004245.1 | NP_001004245.1 | 4 | 606 | ||
| GenBank mRNA | BC079093.1 | - | 4 | 606 | 7123905 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esam| Gene Symbol: Esam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004245.1 | NP_001004245.1 | 4 | 606 | ||
| GenBank mRNA | BC079093.1 | - | 4 | 606 | 7123905 | |