siRNA Details
| Gene Symbol: Olr1252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000454.1 | NP_001000454.1 | 1 | 87 | ||
| Gene Symbol: LOC100912507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762440.1 | XP_008760662.1 | 1 | 87 | ||
| Gene Symbol: LOC103692018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762660.1 | XP_008760882.1 | 1 | 87 | ||

Olr1252
Genomic Map