siRNA Details
| Gene Symbol: Olr1265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000457.1 | NP_001000457.1 | 1 | 788 | ||
| Gene Symbol: LOC100909989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242845.1 | XP_006242907.1 | 2 | 788 | ||
| Gene Symbol: LOC100910111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749184.2 | XP_003749232.1 | 1 | 809 | ||

Olr1265
Genomic Map