siRNA Details
| Gene Symbol: Sorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053519.1 | NP_445971.1 | 534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sorl1| Gene Symbol: Sorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053519.1 | NP_445971.1 | 534 | |||