siRNA Details
| Gene Symbol: Tecta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106814.1 | NP_001100284.1 | 19 | 6136 | ||
| XM_008766130.1 | XP_008764352.1 | 18 | 7477 | |||
| XM_008766131.1 | XP_008764353.1 | 18 | 7462 | |||
| XM_008766132.1 | XP_008764354.1 | 14 | 5042 | |||

Tecta
Genomic Map