siRNA Details
| Gene Symbol: Rnf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113748.1 | NP_001107220.1 | 1 | 1254 | ||
| GenBank mRNA | BC158712.1 | - | 1 | 1254 | 9035415 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf26| Gene Symbol: Rnf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113748.1 | NP_001107220.1 | 1 | 1254 | ||
| GenBank mRNA | BC158712.1 | - | 1 | 1254 | 9035415 | |