siRNA Details
| Gene Symbol: Abcg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106816.1 | NP_001100286.1 | 9, 10 | 1402 | ||
| XM_006242915.2 | XP_006242977.1 | 9, 10 | 7134 | |||
| XM_006242916.2 | XP_006242978.1 | 9, 10 | 1492 | |||

Abcg4
Genomic Map