siRNA Details
| Gene Symbol: Abcg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106816.1 | NP_001100286.1 | 13 | 1912 | ||
| XM_006242915.2 | XP_006242977.1 | 13 | 7632 | |||
| XM_006242916.2 | XP_006242978.1 | 13 | 1990 | |||
| GenBank mRNA | BC089097.1 | - | 1 | 18 | ||

Abcg4
Genomic Map