siRNA Details
| Gene Symbol: Dpagt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199388.1 | NP_955420.1 | 6, 7 | 1047 | ||
| XM_006242921.2 | XP_006242983.1 | 6, 7 | 1398 | |||
| GenBank mRNA | BC063184.1 | - | 6, 7 | 1047 | 6887995 | |
| FQ209511.1 | - | 4, 5 | 486 | |||
| FQ213783.1 | - | 6, 7 | 1124 | |||

Dpagt1
Genomic Map