siRNA Details
| Gene Symbol: Cep164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766182.1 | XP_008764404.1 | 29 | 4009 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep164| Gene Symbol: Cep164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766182.1 | XP_008764404.1 | 29 | 4009 | ||