siRNA Details
| Gene Symbol: Bud13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025277.1 | NP_001020448.1 | 3 | 307 | ||
| GenBank mRNA | BC097995.1 | - | 3 | 307 | 7446660 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bud13| Gene Symbol: Bud13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025277.1 | NP_001020448.1 | 3 | 307 | ||
| GenBank mRNA | BC097995.1 | - | 3 | 307 | 7446660 | |