siRNA Details
| Gene Symbol: Ttc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004246.2 | NP_001004246.1 | 16 | 1492 | ||
| XM_006243004.2 | XP_006243066.1 | 17 | 2305 | |||
| XM_006243005.2 | XP_006243067.1 | 17 | 1560 | |||
| XM_008766196.1 | XP_008764418.1 | 17 | 1707 | |||
| GenBank mRNA | BC079409.1 | - | 16 | 1494 | 7113087 | |

Ttc12
Genomic Map