siRNA Details
Gene Symbol: Etfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009668.1 | NP_001009668.1 | 10 | 923 | ||
GenBank mRNA | BC088412.1 | - | 10 | 923 | 7108329 | |
FQ219339.1 | - | 10 | 947 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Etfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009668.1 | NP_001009668.1 | 10 | 923 | ||
GenBank mRNA | BC088412.1 | - | 10 | 923 | 7108329 | |
FQ219339.1 | - | 10 | 947 |