siRNA Details
| Gene Symbol: Etfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009668.1 | NP_001009668.1 | 3 | 319 | ||
| GenBank mRNA | BC088412.1 | - | 3 | 319 | 7108329 | |
| FQ219339.1 | - | 3 | 342 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etfa| Gene Symbol: Etfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009668.1 | NP_001009668.1 | 3 | 319 | ||
| GenBank mRNA | BC088412.1 | - | 3 | 319 | 7108329 | |
| FQ219339.1 | - | 3 | 342 | |||