siRNA Details
| Gene Symbol: Pstpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106824.2 | NP_001100294.2 | 2 | 208 | ||
| XM_006243113.2 | XP_006243175.1 | 2 | 531 | |||
| XM_008766223.1 | XP_008764445.1 | 2 | 391 | |||
| GenBank mRNA | BC158859.1 | - | 2 | 208 | 7930643 | |

Pstpip1
Genomic Map