siRNA Details
| Gene Symbol: Ubl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004247.1 | NP_001004247.1 | 2 | 227 | ||
| XM_006243155.2 | XP_006243217.1 | 2 | 195 | |||
| XM_006243156.2 | XP_006243218.1 | 2 | 247 | |||
| GenBank mRNA | BC079221.1 | - | 2 | 227 | 7131980 | |
| FQ209768.1 | - | 1 | 184 | |||
| FQ212102.1 | - | 2 | 180 | |||

Ubl7
Genomic Map