siRNA Details
| Gene Symbol: Arih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013108.3 | NP_001013126.2 | 12 | 1508 | ||
| GenBank mRNA | AW141789.1 | - | 2 | 95 | ||
| BC083881.1 | - | 10 | 1393 | 7131418 | ||
| DN956151.1 | - | 9 | 670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arih1| Gene Symbol: Arih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013108.3 | NP_001013126.2 | 12 | 1508 | ||
| GenBank mRNA | AW141789.1 | - | 2 | 95 | ||
| BC083881.1 | - | 10 | 1393 | 7131418 | ||
| DN956151.1 | - | 9 | 670 | |||