siRNA Details
| Gene Symbol: Arih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013108.3 | NP_001013126.2 | 11 | 1457 | ||
| GenBank mRNA | AW141789.1 | - | 1 | 44 | ||
| BC083881.1 | - | 9 | 1342 | 7131418 | ||
| DN956151.1 | - | 8 | 619 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019927, AK087802, BC057680, CK633971, BC038034, and others.

Arih1
Genomic Map