siRNA Details
| Gene Symbol: Gramd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243197.2 | XP_006243259.1 | 6 | 681 | ||
| XM_006243198.2 | XP_006243260.1 | 7 | 1042 | |||
| XM_006243199.2 | XP_006243261.1 | 6 | 1090 | |||
| XM_006243200.2 | XP_006243262.1 | 6 | 1430 | |||
| XM_006243201.2 | XP_006243263.1 | 6 | 2891 | |||
| XM_008766374.1 | XP_008764596.1 | 6 | 561 | |||
| XM_217153.7 | XP_217153.5 | 6 | 480 | |||

Gramd2
Genomic Map