siRNA Details
| Gene Symbol: Vwa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007663.1 | NP_001007664.1 | 9, 10 | 1481 | ||
| XM_008766238.1 | XP_008764460.1 | 10, 11 | 1783 | |||
| XM_008766239.1 | XP_008764461.1 | 11, 12 | 1735 | |||
| GenBank mRNA | BC082006.1 | - | 9, 10 | 1481 | 7131624 | |
| FQ231531.1 | - | 5, 6 | 561 | |||

Vwa9
Genomic Map