siRNA Details
| Gene Symbol: Clpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007803.1 | NP_001007804.1 | 8 | 982 | ||
| XM_006243250.1 | XP_006243312.1 | 7 | 1185 | |||
| GenBank mRNA | BC085867.1 | - | 8 | 1108 | 7190926 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clpx| Gene Symbol: Clpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007803.1 | NP_001007804.1 | 8 | 982 | ||
| XM_006243250.1 | XP_006243312.1 | 7 | 1185 | |||
| GenBank mRNA | BC085867.1 | - | 8 | 1108 | 7190926 | |