siRNA Details
| Gene Symbol: Lactb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106833.1 | NP_001100303.1 | 4 | 948 | ||
| XM_008766242.1 | XP_008764464.1 | 4 | 931 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lactb| Gene Symbol: Lactb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106833.1 | NP_001100303.1 | 4 | 948 | ||
| XM_008766242.1 | XP_008764464.1 | 4 | 931 | |||