siRNA Details
Gene Symbol: Lactb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106833.1 | NP_001100303.1 | 5 | 1093 | ||
GenBank mRNA | FQ230750.1 | - | 2 | 82 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lactb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106833.1 | NP_001100303.1 | 5 | 1093 | ||
GenBank mRNA | FQ230750.1 | - | 2 | 82 |