siRNA Details
| Gene Symbol: Tmod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011997.1 | NP_001011997.1 | 8 | 906 | ||
| GenBank mRNA | BC079290.1 | - | 8 | 906 | 7134366 | |
| FQ229312.1 | - | 3 | 340 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmod3| Gene Symbol: Tmod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011997.1 | NP_001011997.1 | 8 | 906 | ||
| GenBank mRNA | BC079290.1 | - | 8 | 906 | 7134366 | |
| FQ229312.1 | - | 3 | 340 | |||