siRNA Details
| Gene Symbol: Senp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106842.2 | NP_001100312.2 | 11 | 1678 | ||
| XM_006243436.1 | XP_006243498.1 | 10 | 2033 | |||
| XM_006243437.1 | XP_006243499.1 | 9 | 1337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Senp6| Gene Symbol: Senp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106842.2 | NP_001100312.2 | 11 | 1678 | ||
| XM_006243436.1 | XP_006243498.1 | 10 | 2033 | |||
| XM_006243437.1 | XP_006243499.1 | 9 | 1337 | |||